(R)-1,3-Dimethylpiperazin-2-one ≥98%

Supplier: Ambeed
Warning

A526955-1G A526955-250MG
77622-352EA 415.09 USD
77622-352 77622-354
(R)-1,3-Dimethylpiperazin-2-one ≥98%
(R)-1,3-Dimethylpiperazin-2-one

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.1723 g/mol
MDL Number: MFCD22201139
CAS Number: 1240300-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR