N-Methyl-2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide ≥96%
Supplier: Ambeed
|
Danger
|
A535037-100MG
A535037-1G
A535037-250MG
A535037-5G
77886-337EA
39
USD
77886-337
77886-338
77886-339
77886-340
N-Methyl-2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide ≥96%
N-Methyl-2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide
Precaución: for Research Use Only
Formula:
C₁₅H₂₂BNO₃ |
MDL Number:
MFCD17015826 CAS Number: 1036761-96-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...