2-(Bromomethyl)oxetane ≥95%

Supplier: Ambeed
Danger

A526525-1G A526525-250MG
77886-326EA 536 USD
77886-326 77886-327
2-(Bromomethyl)oxetane ≥95%
2-(Bromomethyl)oxetane

Precaución: for Research Use Only

Formula: C₄H₇BrO
MDL Number: MFCD08544405
CAS Number: 939759-23-6
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR