meta-Topolin ≥98%

Supplier: Ambeed
Warning

Synonyms: mT, 3-(((9H-Purin-6-yl)amino)methyl)phenol, 6-(3-Hydroxybenzylamino)purine

A526168-100MG A526168-250MG A526168-1G
77617-244EA 49.44 USD
77617-244 77617-246 77747-074
meta-Topolin ≥98%
meta-Topolin

Precaución: for Research Use Only

Formula: C₁₂H₁₁N₅O
MW: 241.25 g/mol
MDL Number: MFCD03065993
CAS Number: 75737-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR