2,3-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride ≥95%
Supplier: Ambeed
|
Warning
|
A524500-1MG
A524500-10MG
A524500-50MG
A524500-1G
77613-650EA
9.27
USD
77613-650
77686-356
77686-357
77901-347
2,3-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride ≥95%
2,3-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride
Precaución: for Research Use Only
Formula:
C₂₁H₁₈ClNO₄ |
MDL Number:
MFCD01659688 CAS Number: 13063-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...