Chlorobis(3,5-dimethylphenyl)phosphine ≥97%

Supplier: Ambeed
Danger

Synonyms: Bis(3,5-dimethylphenyl)chlorophosphine

A524410-1G A524410-5G A524410-100MG A524410-250MG
77602-242EA 188.6 USD
77602-242 77627-064 77878-308 77878-309
Chlorobis(3,5-dimethylphenyl)phosphine ≥97%
Chlorobis(3,5-dimethylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClP
MDL Number: MFCD01630841
CAS Number: 74289-57-9
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR