2-Bromo-6-fluoro-nicotinic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-fluoronicotinic acid

A524386-100MG A524386-250MG
77891-061EA 72 USD
77891-061 77891-062
2-Bromo-6-fluoro-nicotinic acid ≥95%
2-Bromo-6-fluoro-nicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MDL Number: MFCD13185798
CAS Number: 1214332-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR