(5-Fluoro-2-methylpyridin-4-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A525514-5G A525514-25G A525514-100MG A525514-250MG
77613-654EA 1595.47 USD
77613-654 77684-364 77684-362 77684-363
(5-Fluoro-2-methylpyridin-4-yl)boronic acid ≥98%
(5-Fluoro-2-methylpyridin-4-yl)boronic acid

Precaución: For research use only.

Formula: C₆H₇BFNO₂
MW: 154.94 g/mol
MDL Number: MFCD07374988
CAS Number: 1310404-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR