5-Chloro-2,3-diaminopyridine ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloropyridine-2,3-diyldiamine, 5-Chloropyridine-2,3-diamine, 2,3-Diamino-5-chloropyridine

A524276-1G A524276-5G
77684-358EA 18 USD
77684-358 77684-359
5-Chloro-2,3-diaminopyridine ≥98%
5-Chloro-2,3-diaminopyridine

Precaución: For research use only.

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD01230986
CAS Number: 25710-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR