Spiro[2.3]hexane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A523965-1G A523965-100MG A523965-250MG
77572-668EA 237.93 USD
77572-668 77686-354 77686-355
Spiro[2.3]hexane-1-carboxylic acid ≥98%
Spiro[2.3]hexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.15 g/mol
MDL Number: MFCD04038994
CAS Number: 17202-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR