2-(Methylamino)benzonitrile ≥98%

Supplier: Ambeed
Warning

A523917-1G A523917-250MG
77882-278EA 60 USD
77882-278 77882-279
2-(Methylamino)benzonitrile ≥98%
2-(Methylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD11155692
CAS Number: 17583-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR