Acetylcholine bromide ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetoxy-N,N,N-trimethylethanaminium bromide, (2-Acetoxyethyl)trimethylammonium bromide

A516175-1G A516175-100G A516175-25G A516175-5G
77602-210EA 5.13 USD
77602-210 77627-032 77627-034 77627-036
Acetylcholine bromide ≥98%
Acetylcholine bromide

Precaución: for Research Use Only

Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Br
MW: 226.11 g/mol
MDL Number: MFCD00011814
CAS Number: 66-23-9
Merck Index: 13,00088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR