2,5-Dimethoxybenzenesulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A516023-1G A516023-5G
77747-059EA 17 USD
77747-059 77747-060
2,5-Dimethoxybenzenesulfonyl chloride ≥98%
2,5-Dimethoxybenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₈H₉ClO₄S
MDL Number: MFCD00024872
CAS Number: 1483-28-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR