tert-Butyl 4,7,10,13-tetraoxahexadec-15-yn-1-oate ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propargyl-PEG4-Boc
A515892-1G
A515892-250MG
77749-064EA
154
USD
77749-064
77749-065
tert-Butyl 4,7,10,13-tetraoxahexadec-15-yn-1-oate ≥97%
tert-Butyl 4,7,10,13-tetraoxahexadec-15-yn-1-oate
Precaución: for Research Use Only
Formula:
C₁₆H₂₈O₆ MW: 316.39 g/mol |
MDL Number:
MFCD22683285 CAS Number: 1355197-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...