((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) dimethanesulfonate ≥97%

Supplier: Ambeed
Warning

Synonyms: Ms-PEG4-Ms, 1,11-Bis(methanesulphonyloxy)-3,6,9-trioxandecane

A523514-1G A523514-5G
77749-090EA 53 USD
77749-090 77749-091
((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) dimethanesulfonate ≥97%
((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) dimethanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₉S₂
MW: 350.40 g/mol
MDL Number: MFCD04039441
CAS Number: 55400-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR