6-Bromo-2-iodopyridin-3-amine ≥95%

Supplier: Ambeed
Warning

A515721-100MG A515721-250MG
77884-293EA 22 USD
77884-293 77884-294
6-Bromo-2-iodopyridin-3-amine ≥95%
6-Bromo-2-iodopyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MDL Number: MFCD12963531
CAS Number: 915006-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR