cis-3-(Aminomethyl)cyclobutanol hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A515699-250MG
A515699-1G
A515699-100MG
A515699-5G
77613-622EA
110.21
USD
77613-622
77613-620
77684-340
77684-341
cis-3-(Aminomethyl)cyclobutanol hydrochloride ≥98%
cis-3-(Aminomethyl)cyclobutanol hydrochloride
Precaución: For research use only.
Formula:
C₅H₁₂ClNO |
MDL Number:
MFCD22421655 CAS Number: 1400744-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...