Tris(4-(dimethylamino)phenyl)methanol ≥98%

Supplier: Ambeed
Warning

A515507-100MG A515507-1G A515507-250MG A515507-5G
77872-860EA 72 USD
77872-860 77872-861 77872-862 77872-863
Tris(4-(dimethylamino)phenyl)methanol ≥98%
Tris(4-(dimethylamino)phenyl)methanol

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₃O
MDL Number: MFCD00071920
CAS Number: 467-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR