Dextran sulfate sodium salt, M.W. 40000

Supplier: Ambeed
Warning

A523162-25G A523162-100G A523162-10G A523162-500G A523162-5G
77594-622EA 72.78 USD
77594-622 77594-618 77594-620 77602-238 77602-240
Dextran sulfate sodium salt, M.W. 40000
Dextran sulphate sodium salt

Precaución: for Research Use Only

Formula: (C₆H₇Na₃O₁₄S₃)n
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR