(S)-Methyl 2-((S)-2-(((benzyloxy)carbonyl)amino)-3-methylbutanamido)-3-(4-hydroxyphenyl)propanoate 98+%

Supplier: Ambeed
Warning

A523120-1G A523120-5G A523120-100MG A523120-250MG
77669-316EA 24 USD
77669-316 77872-884 77872-882 77872-883
(S)-Methyl 2-((S)-2-(((benzyloxy)carbonyl)amino)-3-methylbutanamido)-3-(4-hydroxyphenyl)propanoate 98+%
(S)-Methyl 2-((S)-2-(((benzyloxy)carbonyl)amino)-3-methylbutanamido)-3-(4-hydroxyphenyl)propanoate

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₂O₆
MDL Number: MFCD00020191
CAS Number: 15149-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR