Allyl a-D-glucopyranoside ≥97%

Supplier: Ambeed
Warning

A515383-1G A515383-250MG
77878-300EA 212 USD
77878-300 77878-301
Allyl a-D-glucopyranoside ≥97%
Allyl a-D-glucopyranoside

Precaución: for Research Use Only

Formula: C₉H₁₆O₆
MDL Number: MFCD04973540
CAS Number: 7464-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR