N-(tert-Butoxycarbonyl)-D-threonine methyl ester ≥98%

Supplier: Ambeed
Warning

A522776-1G A522776-5G
77747-067EA 101 USD
77747-067 77747-068
N-(tert-Butoxycarbonyl)-D-threonine methyl ester ≥98%
N-(tert-Butoxycarbonyl)-D-threonine methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₅
MDL Number: MFCD01073763
CAS Number: 96099-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR