1-(4-Fluoro-3-(trifluoromethyl)phenyl)-3-(5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl)urea 98+%

Supplier: Ambeed
Warning

Synonyms: ML216

A522753-10MG A522753-1MG A522753-5MG
77617-230EA 43.26 USD
77617-230 77617-232 77617-234
1-(4-Fluoro-3-(trifluoromethyl)phenyl)-3-(5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl)urea 98+%
1-(4-Fluoro-3-(trifluoromethyl)phenyl)-3-(5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl)urea

Precaución: for Research Use Only

Formula: C₁₅H₉F₄N₅OS
MW: 383.32 g/mol
MDL Number: MFCD24849400
CAS Number: 1430213-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR