(R)-tert-Butyl (3-hydroxy-3-methylbutan-2-yl)carbamate ≥95%

Supplier: Ambeed
Warning

A522688-1G A522688-250MG
77673-396EA 176 USD
77673-396 77882-276
(R)-tert-Butyl (3-hydroxy-3-methylbutan-2-yl)carbamate ≥95%
(R)-tert-Butyl (3-hydroxy-3-methylbutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₃
MDL Number: MFCD18831421
CAS Number: 186466-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR