(S)-2-(2-(2-(2-Aminoacetamido)acetamido)-3-phenylpropanamido)acetic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Gly-Gly-Phe-Gly

A514392-250MG A514392-1G A514392-100MG
77617-204EA 181.28 USD
77617-204 77749-061 77749-060
(S)-2-(2-(2-(2-Aminoacetamido)acetamido)-3-phenylpropanamido)acetic acid ≥97%
(S)-2-(2-(2-(2-Aminoacetamido)acetamido)-3-phenylpropanamido)acetic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₄O₅
CAS Number: 200427-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR