3-(Benzyloxy)propanoic acid ≥96%

Supplier: Ambeed
Warning

A522087-1G A522087-25G A522087-5G A522087-250MG
77613-642EA 43.26 USD
77613-642 77613-644 77613-646 77673-390
3-(Benzyloxy)propanoic acid ≥96%
3-(Benzyloxy)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD09951713
CAS Number: 27912-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR