2-Benzyloxy-3-pyridineboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzyloxy)pyridine-3-boronic acid, (2-(Benzyloxy)pyridin-3-yl)boronic acid

A514185-100MG A514185-250MG
77884-289EA 44 USD
77884-289 77884-290
2-Benzyloxy-3-pyridineboronic acid ≥98%
2-Benzyloxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂BNO₃
MDL Number: MFCD06798264
CAS Number: 1072952-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR