(3-Methoxyphenyl)acetyl chloride ≥97%

Supplier: Ambeed
Danger

Synonyms: 2-(3-Methoxyphenyl)acetyl chloride

A514102-1G A514102-5G
77617-200EA 31.93 USD
77617-200 77617-202
(3-Methoxyphenyl)acetyl chloride ≥97%
(3-Methoxyphenyl)acetyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.6196 g/mol
MDL Number: MFCD00012244
CAS Number: 6834-42-0
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR