2,4-Dibromonicotinonitrile ≥95%

Supplier: Ambeed
Danger

A521834-1G A521834-250MG
77891-055EA 115 USD
77891-055 77891-056
2,4-Dibromonicotinonitrile ≥95%
2,4-Dibromonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂Br₂N₂
MDL Number: MFCD22041881
CAS Number: 1152617-14-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR