(R)-2-(Boc-aminomethyl)morpholine ≥98%

Supplier: Ambeed
Warning

A513990-250MG A513990-100MG
77669-282EA 57 USD
77669-282 77872-856
(R)-2-(Boc-aminomethyl)morpholine ≥98%
(R)-2-(Boc-aminomethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD12545850
CAS Number: 186202-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR