Potassium 4-(methylsulfonyl)phenyltrifluoroborate ≥98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(4-(methylsulphonyl)phenyl)borate

A521750-1G A521750-250MG A521750-5G
77669-308EA 65 USD
77669-308 77880-121 77880-122
Potassium 4-(methylsulfonyl)phenyltrifluoroborate ≥98%
Potassium 4-(methylsulphonyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO₂S
MW: 262.09 g/mol
MDL Number: MFCD04115738
CAS Number: 850623-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR