(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((acetamidomethyl)thio)butanoic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-HoCys(Acm)-OH
A521634-100MG
A521634-1G
A521634-250MG
77901-084EA
80
USD
77901-084
77901-994
77901-479
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((acetamidomethyl)thio)butanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((acetamidomethyl)thio)butanoic acid
Precaución: for Research Use Only
Formula:
C₂₂H₂₄N₂O₅S MW: 428.50 g/mol |
MDL Number:
MFCD03788052 CAS Number: 150281-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...