tert-Butyl 6-hydroxyhexanoate ≥95%

Supplier: Ambeed
Warning

A521268-5G A521268-250MG A521268-1G
77572-652EA 303 USD
77572-652 77594-616 77602-230
tert-Butyl 6-hydroxyhexanoate ≥95%
tert-Butyl 6-hydroxyhexanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₃
MW: 188.26 g/mol
MDL Number: MFCD09952156
CAS Number: 73839-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR