(2S,2'S,3R,3'R,4R,4'R,5R,5'R,6S,6'S)-6,6'-(((2R,3S,4S,5R,6R)-4-Hydroxy-2-(hydroxymethyl)-6-(((4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-yl)oxy)tetrahydro-2H-pyran-3,5-diyl)bis(oxy))bis(2-methyltetrahydro-2H-pyran-3,4,5-triol) ≥98%

Supplier: Ambeed
Warning

A521203-1MG A521203-250MG A521203-5MG A521203-100MG A521203-10MG A521203-50MG
77617-222EA 6.18 USD
77617-222 77617-224 77617-226 77617-218 77617-220 77749-085
(2S,2'S,3R,3'R,4R,4'R,5R,5'R,6S,6'S)-6,6'-(((2R,3S,4S,5R,6R)-4-Hydroxy-2-(hydroxymethyl)-6-(((4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-yl)oxy)tetrahydro-2H-pyran-3,5-diyl)bis(oxy))bis(2-methyltetrahydro-2H-pyran-3,4,5-triol) ≥98%
(2S,2'S,3R,3'R,4R,4'R,5R,5'R,6S,6'S)-6,6'-(((2R,3S,4S,5R,6R)-4-Hydroxy-2-(hydroxymethyl)-6-(((4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]in

Precaución: for Research Use Only

Formula: C₄₅H₇₂O₁₆
MW: 869.04 g/mol
MDL Number: MFCD02094174
CAS Number: 19057-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR