(4-(m-Tolyloxy)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A520996-250MG A520996-5G A520996-1G
77901-201EA 57 USD
77901-201 77901-385 77900-691
(4-(m-Tolyloxy)phenyl)boronic acid ≥98%
(4-(m-Tolyloxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₃
MW: 228.05 g/mol
MDL Number: MFCD28024838
CAS Number: 1029438-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR