Zinc(II) 2-aminoacetate ≥99%

Supplier: Ambeed
Danger

A513062-500G A513062-1000G A513062-100G A513062-25G
77598-540EA 56.38 USD
77598-540 77627-018 77878-296 77878-297
Zinc(II) 2-aminoacetate ≥99%
Zinc(II) 2-aminoacetate

Precaución: for Research Use Only

Formula: C₄H₈N₂O₄Zn
CAS Number: 7214-08-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR