4-Bromo-3-fluoro-N-methylaniline ≥97%

Supplier: Ambeed
Warning

A513035-100MG A513035-250MG
77893-040EA 14 USD
77893-040 77893-041
4-Bromo-3-fluoro-N-methylaniline ≥97%
4-Bromo-3-fluoro-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD09745411
CAS Number: 1233513-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR