(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol ≥98%

Supplier: Ambeed
Danger

A520898-50MG A520898-100MG A520898-250MG
77598-542EA 132.23 USD
77598-542 77617-216 77749-084
(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol ≥98%
(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MW: 113.15 g/mol
MDL Number: MFCD27664959
CAS Number: 1638767-26-6
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR