Mal-amido-PEG4-NHS ≥99%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 19-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-17-oxo-4,7,10,13-tetraoxa-16-azanonadecan-1-oate, Mal-amido-PEG4-NHS ester, Maleimide-PEG4-succinimidyl Ester, Maleimide-PEG4-NHS Ester

A512620-100MG A512620-250MG A512620-5G
77613-614EA 33.19 USD
77613-614 77613-616 77686-337
Mal-amido-PEG4-NHS ≥99%
Mal-amido-PEG4-NHS

Precaución: for Research Use Only

Formula: C₂₂H₃₁N₃O₁₁
MDL Number: MFCD11041113
CAS Number: 756525-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR