Potassium trimethoxy(trifluoromethyl)borate ≥95%

Supplier: Ambeed
Warning

Synonyms: Potassium (Trifluoromethyl)trimethoxyborate

A512477-10G A512477-5G A512477-1G
77572-628EA 53 USD
77572-628 77598-538 77617-194
Potassium trimethoxy(trifluoromethyl)borate ≥95%
Potassium trimethoxy(trifluoromethyl)borate

Precaución: for Research Use Only

Formula: C₄H₉BF₃KO₃
MDL Number: MFCD22124929
CAS Number: 626232-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR