5,6-Dimethoxypyridine-2-carbonitrile ≥98%

Supplier: Ambeed
Danger

A512130-250MG A512130-100MG
77673-360EA 26.37 USD
77673-360 77882-260
5,6-Dimethoxypyridine-2-carbonitrile ≥98%
5,6-Dimethoxypyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O₂
MDL Number: MFCD11857661
CAS Number: 1112851-50-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR