3-Bromo-5-fluorobenzene-1,2-diamine ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A519507-25G A519507-1G
77684-350EA 257 USD
77684-350 77886-319
3-Bromo-5-fluorobenzene-1,2-diamine ≥97%, Ambeed.Inc
3-Bromo-5-fluorobenzene-1,2-diamine

Precaución: For research use only.

Formula: C₆H₆BrFN₂
MDL Number: MFCD08741398
CAS Number: 115440-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR