Cefepime hydrochloride monohydrate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cefepime Dihydrochloride Monohydrate, 1-(((6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)-1-methylpyrrolidin-1-ium chloride hydrochloride hydrate
A519450-1MG
A519450-1G
77602-224EA
5.13
USD
77602-224
77627-050
Cefepime hydrochloride monohydrate ≥98%
Cefepime hydrochloride monohydrate
Precaución: for Research Use Only
Formula:
C₁₉H₂₅ClN₆O₅S₂·HCl·H₂O |
MDL Number:
MFCD01687679 CAS Number: 123171-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...