3-Chloro-2-iodobenzonitrile ≥98%

Supplier: Ambeed
Warning

A511568-100MG A511568-250MG
77749-048EA 6 USD
77749-048 77749-049
3-Chloro-2-iodobenzonitrile ≥98%
3-Chloro-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClIN
MW: 263.4629 g/mol
MDL Number: MFCD18249984
CAS Number: 1239493-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR