Benzophenone-4-maleimide ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Benzoylphenyl)-1H-pyrrole-2,5-dione, 4-(N-Maleimido)benzophenone, Benzophenone-4-maleimide

A511386-100MG A511386-1G A511386-250MG
77872-847EA 106 USD
77872-847 77872-848 77872-849
Benzophenone-4-maleimide ≥97%
Benzophenone-4-maleimide

Precaución: for Research Use Only

Formula: C₁₇H₁₁NO₃
MW: 277.2700 g/mol
MDL Number: MFCD00079465
CAS Number: 92944-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR