2-Fluoro-4-methylpyridine-5-carboxyaldehyde ≥98%

Supplier: Ambeed
Warning

A511377-1G A511377-250MG
77884-284EA 70 USD
77884-284 77884-285
2-Fluoro-4-methylpyridine-5-carboxyaldehyde ≥98%
2-Fluoro-4-methylpyridine-5-carboxyaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD18447598
CAS Number: 1266253-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR