4-Bromo-2-(methylthio)pyrimidine ≥95%

Supplier: Ambeed
Danger

A518759-5G A518759-10G
77617-212EA 29.87 USD
77617-212 77749-074
4-Bromo-2-(methylthio)pyrimidine ≥95%
4-Bromo-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂S
MDL Number: MFCD09878932
CAS Number: 959236-97-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR