(5-Bromo-4-chloro-2-fluoro-3-methylphenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A518517-1G A518517-250MG A518517-5G
77882-270EA 189 USD
77882-270 77882-271 77882-272
(5-Bromo-4-chloro-2-fluoro-3-methylphenyl)boronic acid 98+%
(5-Bromo-4-chloro-2-fluoro-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrClFO₂
MDL Number: MFCD31653900
CAS Number: 2377606-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR