(2-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A517486-1G A517486-250MG A517486-5G
77891-047EA 40 USD
77891-047 77891-048 77891-049
(2-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid ≥98%
(2-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₃
MDL Number: MFCD08276821
CAS Number: 1072951-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR