3,6-Dichloro-5-methyl-1,2,4-triazine ≥98%

Supplier: Ambeed
Warning

A517450-100MG A517450-1G A517450-250MG
77594-612EA 75.85 USD
77594-612 77617-206 77617-208
3,6-Dichloro-5-methyl-1,2,4-triazine ≥98%
3,6-Dichloro-5-methyl-1,2,4-triazine

Precaución: for Research Use Only

Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
MDL Number: MFCD18250145
CAS Number: 132434-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR